Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930453H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252013.1 | NP_001238942.1 | 1 - 2 | 442 | 66 | |
GenBank mRNA | AK015451.1 | - | 1 - 2 | 444 | 66 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
synovial sarcoma, X member A, breakpoint 1 |
Ssxa1 |
Gm1532, Ssxa |
-97.4kb | Anti-sense | Chr. X - 21115816 - 21122088 on NCBI Build GRCm38 |
Downstream | predicted gene 6938 | Gm6938 | EG629024 | 90.5kb | Sense | Chr. X - 21312210 - 21334727 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |