Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035122.2 | NP_001030294.1 | 5 - 6 | 541 | 61 | |
NM_027307.4 | NP_081583.3 | 6 - 7 | 677 | 61 | ||
GenBank mRNA | AK009072.1 | - | 4 - 5 | 348 | 61 | |
AK133865.1 | - | 5 - 6 | 543 | 61 | ||
AK167646.1 | - | 5 - 6 | 518 | 61 | ||
BC011152.1 | - | 5 - 6 | 502 | 4188434 | 61 | |
BC098486.1 | - | 6 - 7 | 673 | 30024953 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |