Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290792.1 | NP_001277721.1 | 6 - 7 | 617 | 88 | |
NM_001290794.1 | NP_001277723.1 | 6 - 7 | 513 | 88 | ||
NM_001290795.1 | NP_001277724.1 | 6 - 7 | 617 | 88 | ||
NM_172372.2 | NP_758960.1 | 6 - 7 | 613 | 88 | ||
GenBank mRNA | AF229635.1 | - | 6 - 7 | 591 | 88 | |
AK018409.1 | - | 6 - 7 | 613 | 88 | ||
AK145616.1 | - | 6 - 7 | 513 | 88 | ||
AK205782.1 | - | 1 - 2 | 78 | 88 | ||
AK208276.1 | - | 1 - 2 | 78 | 88 | ||
BC011479.1 | - | 6 - 7 | 584 | 4035468 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |