Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290716.1 | NP_001277645.1 | 1 - 2 | 198 | 136 | |
GenBank mRNA | AF193862.1 | - | 1 - 2 | 142 | 135 | |
AK021174.1 | - | 1 - 2 | 199 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |