Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290716.1 | NP_001277645.1 | 4 - 5 | 1150 | 73 | |
NM_011514.2 | NP_035644.1 | 4 - 5 | 1169 | 73 | ||
GenBank mRNA | AF019969.1 | - | 4 - 5 | 995 | 73 | |
AF193861.1 | - | 4 - 5 | 991 | 73 | ||
AF193862.1 | - | 4 - 5 | 1094 | 73 | ||
AK088405.1 | - | 4 - 5 | 986 | 73 | ||
AK169389.1 | - | 4 - 5 | 977 | 73 | ||
AK169592.1 | - | 4 - 5 | 1139 | 73 | ||
BC023860.1 | - | 4 - 5 | 1046 | 5352230 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |