Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172493.2 | NP_766081.1 | 16 - 17 | 2169 | 121 | |
GenBank mRNA | AB713187.1 | - | 17 - 18 | 1964 | 121 | |
AK083254.1 | - | 9 - 10 | 1101 | 121 | ||
AK088648.1 | - | 16 - 17 | 2103 | 121 | ||
AK153986.1 | - | 16 - 17 | 2157 | 121 | ||
AY312280.1 | - | 16 - 17 | 2106 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |