Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145146.2 | NP_660128.2 | 9 - 10 | 1241 | 134 | |
GenBank mRNA | AK143987.1 | - | 9 - 10 | 1242 | 134 | |
BC026681.1 | - | 9 - 10 | 1159 | 134 | ||
BC100597.1 | - | 9 - 10 | 1241 | 6767654 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |