Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289776.1 | NP_001276705.1 | 2 - 3 | 860 | 68 | |
NM_001289777.1 | NP_001276706.1 | 3 - 4 | 685 | 68 | ||
GenBank mRNA | AK013185.1 | - | 3 - 4 | 686 | 68 | |
BC058259.1 | - | 2 - 3 | 579 | 6469137 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |