Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK167967.1 | - | 3 - 4 | 288 | 77 | |
AK168338.1 | - | 3 - 4 | 288 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |