Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271476.1 | NP_001258405.1 | 1 - 2 | 175 | 66 | |
NM_001271478.1 | NP_001258407.1 | 1 - 2 | 175 | 66 | ||
GenBank mRNA | AB084140.1 | - | 1 - 2 | 175 | 66 | |
AB084141.1 | - | 1 - 2 | 175 | 66 | ||
AF392055.1 | - | 1 - 2 | 101 | 66 | ||
AK039953.1 | - | 1 - 2 | 175 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |