Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134127.1 | NP_598888.1 | 2 - 3 | 355 | 62 | |
GenBank mRNA | AF233645.1 | - | 2 - 3 | 355 | 62 | |
BC021377.1 | - | 2 - 3 | 346 | 5050617 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |