Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177823.4 | NP_808491.2 | 1 - 2 | 168 | 109 | |
GenBank mRNA | AK040572.1 | - | 1 - 2 | 128 | 109 | |
AK041690.1 | - | 1 - 2 | 135 | 109 | ||
BC027561.1 | - | 1 - 2 | 154 | 109 | ||
BY227020.1 | - | 1 - 2 | 168 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |