Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177823.4 | NP_808491.2 | 3 - 4 | 405 | 66 | |
GenBank mRNA | AK040572.1 | - | 3 - 4 | 365 | 66 | |
AK041690.1 | - | 3 - 4 | 372 | 66 | ||
AK049106.1 | - | 2 - 3 | 305 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |