Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026179.2 | NP_080455.1 | 3 - 4 | 686 | 70 | |
GenBank mRNA | AK004873.1 | - | 3 - 4 | 572 | 70 | |
AK007421.1 | - | 3 - 4 | 568 | 70 | ||
AK028453.1 | - | 3 - 4 | 572 | 70 | ||
AK050377.1 | - | 3 - 4 | 571 | 70 | ||
AK152717.1 | - | 4 - 5 | 620 | 70 | ||
BC037063.1 | - | 3 - 4 | 568 | 4985992 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |