Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037761.2 | NP_001032850.1 | 1 - 2 | 131 | 83 | |
NM_009798.4 | NP_033928.1 | 1 - 2 | 131 | 83 | ||
GenBank mRNA | AK153367.1 | - | 1 - 2 | 62 | 83 | |
AK153394.1 | - | 1 - 2 | 62 | 83 | ||
AK156778.1 | - | 1 - 2 | 100 | 83 | ||
AK159390.1 | - | 1 - 2 | 106 | 82 | ||
AK160082.1 | - | 1 - 2 | 106 | 83 | ||
AK168738.1 | - | 1 - 2 | 59 | 83 | ||
BC002053.1 | - | 1 - 2 | 73 | 3590980 | 83 | |
BY124858.1 | - | 1 - 2 | 120 | 83 | ||
U10406.1 | - | 1 - 2 | 51 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |