Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crocc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145958.1 | NP_001139430.1 | 25 - 26 | 3868 | 59 | |
NM_172122.2 | NP_742120.2 | 26 - 27 | 4121 | 59 | ||
GenBank mRNA | AF527975.1 | - | 26 - 27 | 4121 | 59 | |
BC034536.1 | - | 3 - 4 | 1026 | 59 | ||
BC036729.1 | - | 11 - 12 | 1763 | 59 | ||
BC054054.1 | - | 24 - 25 | 3849 | 6394697 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |