Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009951.1 | NP_001009951.1 | 6 - 7 | 741 | 92 | |
GenBank mRNA | AK220415.1 | - | 5 - 6 | 697 | 92 | |
BC088983.1 | - | 6 - 7 | 741 | 5700628 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |