Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172338.2 | NP_758841.1 | 1 - 2 | 55 | 71 | |
GenBank mRNA | AK028692.1 | - | 1 - 2 | 60 | 71 | |
AK051048.1 | - | 1 - 2 | 72 | |||
AK122407.1 | - | 302 | 71 | |||
AK154946.1 | - | 76 | 71 | |||
AK164023.1 | - | 1 - 2 | 71 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |