Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026818.2 | NP_081094.2 | 6 - 7 | 1017 | 96 | |
GenBank mRNA | BC006187.1 | - | 6 - 7 | 677 | 96 | |
BC117911.2 | - | 2 - 3 | 96 | |||
BC117912.2 | - | 2 - 3 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |