Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113346.1 | NP_001106817.1 | 3 - 4 | 661 | 56 | |
NM_001286450.1 | NP_001273379.1 | 3 - 4 | 809 | 56 | ||
NM_145596.4 | NP_663571.2 | 3 - 4 | 809 | 56 | ||
GenBank mRNA | AK083473.1 | - | 3 - 4 | 811 | 56 | |
AK089074.1 | - | 3 - 4 | 791 | 56 | ||
AK160058.1 | - | 3 - 4 | 658 | 56 | ||
BC038221.1 | - | 3 - 4 | 559 | 5321913 | 56 | |
CN666280.1 | - | 1 - 2 | 179 | 56 | ||
CV562594.1 | - | 3 - 4 | 567 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |