Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027560.1 | NP_081836.1 | 2 - 3 | 655 | 62 | |
GenBank mRNA | AK014582.1 | - | 2 - 3 | 655 | 62 | |
AK046754.1 | - | 2 - 3 | 365 | 62 | ||
AK135740.1 | - | 2 - 3 | 471 | 62 | ||
AK137175.1 | - | 2 - 3 | 472 | 62 | ||
AK144323.1 | - | 3 - 4 | 389 | 62 | ||
BC036384.1 | - | 2 - 3 | 453 | 1067080 | 62 | |
BC094060.1 | - | 1 - 2 | 535 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |