Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027560.1 | NP_081836.1 | 3 - 4 | 811 | 95 | |
GenBank mRNA | AK014582.1 | - | 3 - 4 | 811 | 95 | |
AK046754.1 | - | 3 - 4 | 521 | 95 | ||
AK135740.1 | - | 3 - 4 | 627 | 95 | ||
AK137175.1 | - | 3 - 4 | 628 | 95 | ||
AK144323.1 | - | 4 - 5 | 545 | 95 | ||
BC036384.1 | - | 3 - 4 | 609 | 1067080 | 95 | |
BC094060.1 | - | 2 - 3 | 691 | 95 | ||
DQ861995.1 | - | 3 - 4 | 475 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |