Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027560.1 | NP_081836.1 | 4 - 5 | 911 | 69 | |
GenBank mRNA | AK014582.1 | - | 4 - 5 | 911 | 69 | |
AK046754.1 | - | 4 - 5 | 621 | 69 | ||
AK135740.1 | - | 4 - 5 | 727 | 69 | ||
AK137175.1 | - | 4 - 5 | 728 | 69 | ||
AK144323.1 | - | 5 - 6 | 645 | 69 | ||
BC036384.1 | - | 4 - 5 | 709 | 1067080 | 69 | |
BC094060.1 | - | 3 - 4 | 791 | 69 | ||
DQ861995.1 | - | 4 - 5 | 575 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |