Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023217.4 | NP_075706.1 | 1 - 2 | 131 | 66 | |
GenBank mRNA | AJ278829.1 | - | 1 - 2 | 63 | 66 | |
AK003373.1 | - | 1 - 2 | 75 | 66 | ||
AK012658.1 | - | 1 - 2 | 63 | 66 | ||
AK047835.1 | - | 1 - 2 | 75 | 66 | ||
AK049632.1 | - | 1 - 2 | 54 | 66 | ||
BC043041.1 | - | 1 - 2 | 41 | 66 | ||
BC051938.1 | - | 1 - 2 | 60 | 5717365 | 66 | |
BY313315.1 | - | 1 - 2 | 132 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |