Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004062.2 | NP_001004062.1 | 10 - 11 | 1324 | 66 | |
GenBank mRNA | AK129173.1 | - | 10 - 11 | 1321 | 66 | |
BC056137.1 | - | 10 - 11 | 5912 | 66 | ||
BC068139.1 | - | 10 - 11 | 1330 | 66 | ||
BC080308.1 | - | 10 - 11 | 1324 | 5719562 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |