Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133772.2 | NP_598533.2 | 12 - 13 | 905 | 62 | |
GenBank mRNA | AK004835.1 | - | 12 - 13 | 1018 | 62 | |
AK154336.1 | - | 12 - 13 | 906 | 62 | ||
AK157387.1 | - | 11 - 12 | 985 | 62 | ||
AK183440.1 | - | 2 - 3 | 97 | 62 | ||
AK188774.1 | - | 2 - 3 | 97 | 62 | ||
BC002150.1 | - | 12 - 13 | 864 | 3484896 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |