Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277876.1 | NP_001264805.1 | 5 - 6 | 762 | 79 | |
GenBank mRNA | BC014809.1 | - | 5 - 6 | 646 | 3497670 | 79 |
BC024358.1 | - | 5 - 6 | 660 | 4990394 | 79 | |
BY439632.1 | - | 1 - 2 | 54 | 79 | ||
CV560790.1 | - | 5 - 6 | 537 | 79 | ||
X58381.1 | - | 5 - 6 | 638 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |