Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277876.1 | NP_001264805.1 | 6 - 7 | 834 | 104 | |
GenBank mRNA | AA208641.1 | - | 2 - 3 | 101 | 104 | |
BC024358.1 | - | 6 - 7 | 732 | 4990394 | 104 | |
X58381.1 | - | 6 - 7 | 710 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |