Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170430.1 | NP_001163901.1 | 6 - 7 | 894 | 74 | |
NM_001170431.1 | NP_001163902.1 | 6 - 7 | 1029 | 74 | ||
NM_134060.4 | NP_598821.1 | 6 - 7 | 872 | 74 | ||
GenBank mRNA | AK076592.1 | - | 6 - 7 | 867 | 74 | |
AK077208.1 | - | 6 - 7 | 848 | 74 | ||
AK141280.1 | - | 6 - 7 | 1030 | 74 | ||
BC006881.1 | - | 6 - 7 | 886 | 3596363 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |