Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026577.3 | NP_080853.3 | 8 - 9 | 1438 | 72 | |
GenBank mRNA | BC082574.1 | - | 8 - 9 | 1438 | 30550373 | 72 |
CK781880.1 | - | 1 - 2 | 81 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |