Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283028.1 | NP_001269957.1 | 2 - 3 | 387 | 89 | |
NM_001283029.1 | NP_001269958.1 | 2 - 3 | 252 | 89 | ||
NM_146043.4 | NP_666155.1 | 2 - 3 | 312 | 89 | ||
GenBank mRNA | AK088127.1 | - | 2 - 3 | 253 | 89 | |
AK134259.1 | - | 2 - 3 | 275 | 89 | ||
AK168544.1 | - | 2 - 3 | 313 | 89 | ||
BC016517.1 | - | 2 - 3 | 298 | 2654233 | 89 | |
CJ173506.1 | - | 2 - 3 | 371 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |