Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283028.1 | NP_001269957.1 | 4 - 5 | 692 | 66 | |
NM_001283029.1 | NP_001269958.1 | 4 - 5 | 557 | 66 | ||
NM_001283030.1 | NP_001269959.1 | 3 - 4 | 416 | 66 | ||
NM_011462.3 | NP_035592.1 | 2 - 3 | 321 | 66 | ||
NM_146043.4 | NP_666155.1 | 4 - 5 | 617 | 66 | ||
GenBank mRNA | AK088127.1 | - | 4 - 5 | 558 | 66 | |
AK134259.1 | - | 4 - 5 | 580 | 66 | ||
AK135816.1 | - | 2 - 3 | 316 | 66 | ||
AK135847.1 | - | 2 - 3 | 319 | 66 | ||
AK135933.1 | - | 2 - 3 | 319 | 66 | ||
AK136233.1 | - | 2 - 3 | 319 | 66 | ||
AK136251.1 | - | 2 - 3 | 320 | 66 | ||
AK139899.1 | - | 2 - 3 | 318 | 66 | ||
AK162171.1 | - | 2 - 3 | 318 | 66 | ||
AK162195.1 | - | 2 - 3 | 318 | 66 | ||
AK162258.1 | - | 2 - 3 | 315 | 66 | ||
AK163263.1 | - | 2 - 3 | 317 | 66 | ||
AK163293.1 | - | 2 - 3 | 318 | 66 | ||
AK163301.1 | - | 2 - 3 | 322 | 66 | ||
AK163308.1 | - | 2 - 3 | 320 | 66 | ||
AK163382.1 | - | 2 - 3 | 318 | 66 | ||
AK166528.1 | - | 2 - 3 | 321 | 66 | ||
AK166532.1 | - | 2 - 3 | 321 | 66 | ||
AK166580.1 | - | 2 - 3 | 280 | 66 | ||
AK166677.1 | - | 2 - 3 | 317 | 66 | ||
AK168544.1 | - | 4 - 5 | 618 | 66 | ||
BC016517.1 | - | 4 - 5 | 603 | 2654233 | 66 | |
U48972.1 | - | 2 - 3 | 315 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |