Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027123.4 | NP_081399.3 | 5 - 6 | 1903 | 62 | |
GenBank mRNA | AK009264.1 | - | 5 - 6 | 662 | 62 | |
AK032530.1 | - | 5 - 6 | 1986 | 62 | ||
AK033189.1 | - | 5 - 6 | 1904 | 62 | ||
AK036778.1 | - | 5 - 6 | 1986 | 62 | ||
AK148902.1 | - | 5 - 6 | 1894 | 62 | ||
BC042506.1 | - | 5 - 6 | 1977 | 4023412 | 62 | |
BC060688.1 | - | 4 - 5 | 4616 | 6409274 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |