Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025495.3 | NP_079771.1 | 2 - 3 | 320 | 164 | |
GenBank mRNA | AK006234.1 | - | 2 - 3 | 357 | 164 | |
AK012863.1 | - | 2 - 3 | 324 | 164 | ||
AK014919.1 | - | 2 - 3 | 276 | 164 | ||
AK076577.1 | - | 2 - 3 | 358 | 164 | ||
BC094280.1 | - | 2 - 3 | 374 | 164 | ||
BC147207.1 | - | 2 - 3 | 300 | 9056262 | 164 | |
BC147208.1 | - | 2 - 3 | 300 | 9056260 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |