Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038993.3 | NP_001034082.1 | 8 - 9 | 1209 | 134 | |
NM_175201.5 | NP_780410.2 | 7 - 8 | 1239 | 134 | ||
GenBank mRNA | AK147608.1 | - | 8 - 9 | 1266 | 134 | |
AK163346.1 | - | 3 - 4 | 370 | 134 | ||
BC060730.1 | - | 7 - 8 | 1216 | 6408623 | 134 | |
BC062976.1 | - | 7 - 8 | 1153 | 6848803 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |