Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080385.1 | NP_001073854.1 | 4 - 5 | 631 | 66 | |
NM_016760.2 | NP_058040.2 | 4 - 5 | 631 | 66 | ||
GenBank mRNA | CA322069.1 | - | 4 - 5 | 594 | 66 | |
CX243404.1 | - | 4 - 5 | 583 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |