Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165902.1 | NP_001159374.1 | 5 - 6 | 825 | 57 | |
NM_007614.3 | NP_031640.1 | 5 - 6 | 1025 | 57 | ||
GenBank mRNA | AK018515.1 | - | 5 - 6 | 939 | 57 | |
AK035311.1 | - | 5 - 6 | 938 | 57 | ||
AK132398.1 | - | 5 - 6 | 1006 | 57 | ||
AK133656.1 | - | 5 - 6 | 938 | 57 | ||
AK135743.1 | - | 2 - 3 | 492 | 57 | ||
AK197753.1 | - | 1 - 2 | 65 | 57 | ||
AY751549.1 | - | 5 - 6 | 950 | 57 | ||
BC043078.1 | - | 2 - 3 | 427 | 57 | ||
BC043481.1 | - | 1 - 2 | 59 | 57 | ||
BC048153.1 | - | 5 - 6 | 950 | 5709247 | 57 | |
BC053065.1 | - | 5 - 6 | 950 | 5709247 | 57 | |
M90364.1 | - | 5 - 6 | 835 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |