Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165902.1 | NP_001159374.1 | 8 - 9 | 1270 | 91 | |
NM_007614.3 | NP_031640.1 | 8 - 9 | 1470 | 91 | ||
GenBank mRNA | AK018515.1 | - | 8 - 9 | 1384 | 91 | |
AK020013.1 | - | 8 - 9 | 1386 | 91 | ||
AK035311.1 | - | 8 - 9 | 1383 | 91 | ||
AK132398.1 | - | 8 - 9 | 1451 | 91 | ||
AK133656.1 | - | 8 - 9 | 1383 | 91 | ||
AK135743.1 | - | 5 - 6 | 937 | 91 | ||
AK184861.1 | - | 2 - 3 | 120 | 91 | ||
AY751549.1 | - | 8 - 9 | 1395 | 91 | ||
BC043078.1 | - | 5 - 6 | 872 | 91 | ||
BC043481.1 | - | 4 - 5 | 504 | 91 | ||
BC048153.1 | - | 8 - 9 | 1395 | 5709247 | 91 | |
BC053065.1 | - | 8 - 9 | 1395 | 5709247 | 91 | |
M90364.1 | - | 8 - 9 | 1280 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |