Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172121.1 | NP_001165592.1 | 4 - 5 | 496 | 82 | |
NM_001172122.1 | NP_001165593.1 | 3 - 4 | 968 | 82 | ||
NM_001172123.1 | NP_001165594.1 | 3 - 4 | 968 | 82 | ||
NM_001172124.1 | NP_001165595.1 | 3 - 4 | 968 | 82 | ||
NM_001172126.1 | NP_001165597.1 | 3 - 4 | 968 | 82 | ||
NM_178660.4 | NP_848775.2 | 3 - 4 | 968 | 82 | ||
GenBank mRNA | AK030478.1 | - | 3 - 4 | 628 | 82 | |
AK030609.1 | - | 3 - 4 | 602 | 82 | ||
AK032938.1 | - | 3 - 4 | 965 | 82 | ||
AK033005.1 | - | 3 - 4 | 611 | 82 | ||
AK041098.1 | - | 3 - 4 | 970 | 82 | ||
AK046197.1 | - | 4 - 5 | 497 | 82 | ||
AK051776.1 | - | 3 - 4 | 967 | 82 | ||
AK052196.1 | - | 3 - 4 | 949 | 82 | ||
AK081921.1 | - | 3 - 4 | 962 | 82 | ||
AK162457.1 | - | 3 - 4 | 333 | 82 | ||
BC108402.1 | - | 3 - 4 | 942 | 6516420 | 82 | |
BC117841.1 | - | 3 - 4 | 408 | 40093225 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |