Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252513.1 | NP_001239442.1 | 1 - 2 | 150 | 115 | |
GenBank mRNA | AK086576.1 | - | 1 - 2 | 153 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |