Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172693.3 | NP_766281.1 | 7 - 8 | 1349 | 61 | |
GenBank mRNA | AB175680.1 | - | 7 - 8 | 1329 | 61 | |
AK033638.1 | - | 7 - 8 | 1350 | 61 | ||
AK042133.1 | - | 7 - 8 | 1382 | 61 | ||
AK043977.1 | - | 6 - 7 | 1210 | 61 | ||
AK082014.1 | - | 7 - 8 | 1395 | 61 | ||
AK156751.1 | - | 7 - 8 | 1379 | 61 | ||
BC024475.1 | - | 6 - 7 | 1181 | 61 | ||
BC025639.1 | - | 6 - 7 | 1071 | 61 | ||
BC056425.1 | - | 7 - 8 | 1360 | 5716909 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |