Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rp2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290644.1 | NP_001277573.1 | 2 - 3 | 509 | 85 | |
GenBank mRNA | AK033954.1 | - | 2 - 3 | 432 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |