Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010918.2 | NP_035048.3 | 7 - 8 | 544 | 151 | |
GenBank mRNA | AK035582.1 | - | 6 - 7 | 445 | 151 | |
AK144214.1 | - | 7 - 8 | 455 | 151 | ||
BC048414.1 | - | 8 - 9 | 534 | 179 | ||
L04289.2 | - | 7 - 8 | 549 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |