Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028384.1 | NP_082660.1 | 14 - 15 | 1911 | 67 | |
GenBank mRNA | AK038868.1 | - | 4 - 5 | 409 | 67 | |
AW911821.1 | - | 1 - 2 | 162 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |