Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081990 |
BC144974.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081990 |
BC137851.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081990 |
BC132477.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081990 |
AK016425.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081990 |
NM_029401.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124549 |
BC144974.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124549 |
BC137851.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124549 |
BC132477.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124549 |
NM_029401.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306387 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
966468 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
13616 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
940402 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
148053 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
37904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
190149 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
231272 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729935 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
613732 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
243589 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
134558 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
13832 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656195 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
819123 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45986 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
244825 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704313 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798411 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
644037 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
797858 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
883697 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
275478 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
313424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344307 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
AK076815.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
AK147946.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
AK169684.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
BC086668.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
BC110673.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
L21027.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895161 |
NM_016966.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
318784 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647731 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
147422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948600 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
17854 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
55567 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
17149 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
76555 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
36260 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
85312 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P180515 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P161252 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124549 |
BC144974.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124549 |
BC137851.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124549 |
BC132477.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124549 |
NM_029401.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081990 |
BC144974.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081990 |
BC137851.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081990 |
BC132477.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081990 |
AK016425.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081990 |
NM_029401.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P180515 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01002067 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05422905 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05422846 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P161252 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05422879 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P104409 |
BC137851.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P104409 |
BC132477.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P104409 |
BC057376.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P104409 |
AK016425.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P104409 |
NM_029401.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05422879 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P180515 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P161252 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346899 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346894 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346778 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346878 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346595 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346869 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346617 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03204352 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03528591 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03542139 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03296169 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346889 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03949843 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03298792 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346614 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346716 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346879 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346711 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03982444 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346835 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03905185 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346902 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346706 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04094444 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346626 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346570 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346828 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346710 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346707 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03803738 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346743 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346891 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03522411 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346756 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346597 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03459758 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03312792 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346635 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346609 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346903 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346566 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346907 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03296172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346572 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03930049 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03930048 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346584 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346581 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346753 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03928500 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346884 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03982450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346564 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346591 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346821 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03774935 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346713 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346567 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03928504 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03278154 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346846 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346850 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346586 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346825 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03522410 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346701 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03508095 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03520293 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346841 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346600 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346859 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346592 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346742 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346563 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346624 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346909 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346837 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346739 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04095530 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346754 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346782 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03220601 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346844 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346857 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346632 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03250898 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346897 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346875 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03614807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04094439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04094440 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346601 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03441124 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346620 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03948923 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03511556 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03665342 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346615 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03797375 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346580 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03441125 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346790 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03204353 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346750 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346826 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03684020 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346588 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346849 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346862 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03774928 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03774932 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346561 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346864 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346838 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03414918 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346574 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346622 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03928502 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03296175 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346623 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04082372 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346794 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346873 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346866 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346748 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03774929 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346901 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03684019 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23346583 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03592208 |
- |
- |