Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164612.1 | NP_001158084.1 | 5 - 6 | 710 | 100 | |
NM_001164613.1 | NP_001158085.1 | 6 - 7 | 763 | 100 | ||
NM_172613.4 | NP_766201.3 | 5 - 6 | 710 | 100 | ||
GenBank mRNA | AK028776.1 | - | 6 - 7 | 765 | 100 | |
BC048410.1 | - | 5 - 6 | 710 | 6486835 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |