Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007423.4 | NP_031449.3 | 8 - 9 | 1110 | 157 | |
GenBank mRNA | AK010934.1 | - | 8 - 9 | 1086 | 157 | |
AK075972.1 | - | 8 - 9 | 1090 | 157 | ||
AK076053.1 | - | 8 - 9 | 1086 | 157 | ||
AK076197.1 | - | 8 - 9 | 1088 | 157 | ||
AK145975.1 | - | 8 - 9 | 1086 | 157 | ||
AK146513.1 | - | 8 - 9 | 1088 | 157 | ||
AK146524.1 | - | 8 - 9 | 1088 | 157 | ||
AK167549.1 | - | 8 - 9 | 1086 | 157 | ||
AK168426.1 | - | 8 - 9 | 1089 | 157 | ||
AK168574.1 | - | 8 - 9 | 1088 | 157 | ||
AK168576.1 | - | 8 - 9 | 1086 | 157 | ||
AK168751.1 | - | 8 - 9 | 1088 | 157 | ||
AK168818.1 | - | 7 - 8 | 964 | 157 | ||
AK168883.1 | - | 8 - 9 | 1086 | 157 | ||
AK169046.1 | - | 8 - 9 | 1088 | 157 | ||
AK169054.1 | - | 8 - 9 | 1088 | 157 | ||
AK169147.1 | - | 8 - 9 | 1086 | 157 | ||
BC066206.1 | - | 8 - 9 | 1073 | 30469872 | 157 | |
V00743.1 | - | 8 - 9 | 1084 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |