Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177195.3 | NP_796169.2 | 16 - 17 | 1623 | 60 | |
GenBank mRNA | AK029992.1 | - | 17 - 18 | 1853 | 60 | |
EF377342.1 | - | 16 - 17 | 1612 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Mus musculus Probe spans exons |