Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173788.3 | NP_776149.1 | 11 - 12 | 2055 | 78 | |
GenBank mRNA | BC037458.1 | - | 6 - 7 | 511 | 78 | |
BC042470.1 | - | 11 - 12 | 2055 | 5137261 | 78 | |
BC049896.1 | - | 1 - 2 | 152 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |