Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172692.3 | NP_766280.2 | 13 - 14 | 2543 | 101 | |
GenBank mRNA | AK089192.1 | - | 13 - 14 | 2229 | 101 | |
AK158174.1 | - | 11 - 12 | 2914 | 101 | ||
AK173213.1 | - | 13 - 14 | 2529 | 101 | ||
BC031397.1 | - | 11 - 12 | 2546 | 101 | ||
BC034105.1 | - | 10 - 11 | 2294 | 101 | ||
BC056935.1 | - | 13 - 14 | 2066 | 5703478 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |